Key Documents
Z167525
Sigma-Aldrich
Aldrich® Schlenk-type Liebig condenser
male joint: ST/NS 14/20, female joint: ST/NS 14/20
Select a Size
1 EA
$141.00
$141.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$141.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$141.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
label
P
Joint
female joint: ST/NS14/20
male joint: ST/NS14/20
L
100 mm
Related Categories
General description
Replacement part for Aldrich Air-Sensitive glass kit. Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.