Key Documents
Z218707
Clamps
fits flask, 2,000 ‑ 4,000 mL
Synonym(s):
flask clamp, flask lab, lab flask, reaction flasks
Select a Size
1 EA
$851.00
$851.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$851.00
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
$851.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
feature
fits flask 2,000 ‑ 4,000 mL
manufacturer/tradename
Ace Glass 650811
General description
Two-piece, black anodized aluminum clamp has three wing nuts to secure Ace reaction flask to head. Clamps also fit Pyrex reaction kettles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)