Key Documents
Z555800
Ace reaction flask head
capacity 1,000 mL, center joint: ST/NS 24/40, side Joints: ST/NS 24/40 (2)
Synonym(s):
flask lab, lab flask, reaction flasks
Select a Size
1 EA
$954.00
$954.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$954.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$954.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
material
glass
three-neck (in-line)
Joint
center joint: ST/NS24/40
side Joints: ST/NS24/40 (2)
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Ace Glass 651216
capacity
1,000 mL
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)