Key Documents
Z201022
Sigma-Aldrich
Spilfyter® chemical classifier
pkg of 1 ea
Select a Size
1 EA
$229.00
$229.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$229.00
About This Item
UNSPSC Code:
47131905
NACRES:
NB.35
$229.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pkg of 1 ea
General description
Quickly distinguish unknown spilled or stored chemicals. Classifier consists of a test strip with various positioned color indicators that are sensitive to different chemical classes (e.g., pH, oxidizers, fluorides, organics, and chlorides). Strips are sealed against premature exposure by a protective film.
Each kit includes six classifiers, 120 ft PP string, and a laminated color-identification chart. Shipped in an airtight tube.
Legal Information
Spilfyter is a registered trademark of National Packaging Services Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)