Key Documents
Z176966
Scienceware® serum-vial support
Holds 72 x 10-13 mm tubes
Select a Size
1 EA
$53.40
$53.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$53.40
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
$53.40
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
feature
Holds 72 x 10-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18513-0072
L × W × H
9 3/8 in. × 4 3/8 in. × 3 1/2 in.
General description
Lightweight PP. Corrosion-proof. Fits most water baths for elevated temperature incubations. Top shelf height is 41mm (1⁵⁄₈").
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)