Key Documents
Z173177
Sigma-Aldrich
Aldrich® Schlenk-type adapter
Straight with PTFE stopcock on side-arm, male joint: threaded 14/20, female joint: threaded 14/20
Select a Size
1 EA
$304.00
$304.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$304.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$304.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
female joint: threaded14/20
male joint: threaded14/20
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)