Documents
Z165220
Sigma-Aldrich
Cork-borer set
zinc-plated brass set of 6
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
General description
For boring holes in corks and rubber stoppers. They consist of zinc-plated brass tubing with precision ground cutting edges. Thesmooth plastic handles are stamped with a size (3/16, 1/4, 5/16, 3/8, 7/16 and 1/2 in.) designation, which corresponds to the table below
Please note: A punch is included with each set.
Visit the Document Library
Robert M Q Shanks et al.
PloS one, 7(5), e36398-e36398 (2012-05-23)
Serratia marcescens is a common contaminant of contact lens cases and lenses. Hemolytic factors of S. marcescens contribute to the virulence of this opportunistic bacterial pathogen. We took advantage of an observed hyper-hemolytic phenotype of crp mutants to investigate mechanisms
Bruno Pok Man Ngou et al.
Journal of experimental botany, 71(6), 2186-2197 (2020-02-13)
Plant nucleotide-binding domain, leucine-rich repeat receptor (NLR) proteins play important roles in recognition of pathogen-derived effectors. However, the mechanism by which plant NLRs activate immunity is still largely unknown. The paired Arabidopsis NLRs RRS1-R and RPS4, that confer recognition of
Protocols
This protocol covers three methods for the microscopic examination of cell samples.