Key Documents
Z162558
Supelco
Septa for open top caps
for cap size 8 mm, red PTFE/silicone
About This Item
UNSPSC Code:
41122001
material
red PTFE/silicone
packaging
pkg of 100
General description
For use with cut-out top caps and Wheaton vials.
Choose from one of the most recent versions:
Certificates of Analysis (COA)