Key Documents
Z158232
Ace Enlarging Adapter
female joint: ST/NS 29/42, male joint: ST/NS 24/40
Synonym(s):
glass adapter, glass adapters
Select a Size
1 EA
$134.00
$134.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$134.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$134.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
female joint: ST/NS29/42
male joint: ST/NS24/40
manufacturer/tradename
Ace Glass
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)