Key Documents
SYNA252924
Synthware enlarging adapter
top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonym(s):
Synthware A252924, glass connecting adapter
Select a Size
1 EA
$88.00
$88.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$88.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$88.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A252924
Related Categories
General description
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (29/42) is larger that the bottom joint (24/40).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)