Key Documents
Z150762
Sigma-Aldrich
Water aspirator pump
Short swirling
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
L
76 mm
General description
Operates efficiently on both high and low water pressure. Nickel-plated 3/8 in. NPTM water inlet. Ball-type, positive-acting valve in suction tube prevents backflow should pressure be reduced. Max. vacuum is 100 mm Hg (abs).
With serrated tail and side-arm.
Choose from one of the most recent versions:
Certificates of Analysis (COA)