Key Documents
Z150355
Sigma-Aldrich
Aldrich® septum-inlet adapter with stopcock
Plain, joint: ST/NS 24/40, PTFE stopcock
Select a Size
1 EA
$240.00
$240.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$240.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$240.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
PTFE stopcock
Joint
joint: ST/NS24/40
inlet diam.
14 mm
Related Categories
General description
Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)