Key Documents
Z150320
Sigma-Aldrich
Aldrich® septum-inlet adapter, male
Plain, joint: ST/NS 24/40
Select a Size
1 EA
$59.80
$59.80
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$59.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$59.80
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Joint
joint: ST/NS24/40
General description
Inlet diam. 14 mm. Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)