Key Documents
Z136107
Sigma-Aldrich
Scienceware® pipette support rack
holds 50 pipettes vertically in a 5 × 10 arrangement
Select a Size
1 EA
$61.90
$61.90
Estimated to ship onMarch 29, 2025
Select a Size
Change View
1 EA
$61.90
About This Item
UNSPSC Code:
41122801
NACRES:
NB.01
$61.90
Estimated to ship onMarch 29, 2025
material
polypropylene
feature
Holds 50x pipettes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18953-0000
L × W × H
8.375 cm × 4.5 cm × 8.75 cm
General description
Molded PP rack is autoclavable (121°C). Holds 50 pipettes vertically in a 5 × 10 arrangement. Holes in top and middle deck are 16 mm diam., and holes in lower deck are tapered from 16 to 6 mm diam. Lower deck prevents pipette tips from touching table beneath.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)