Key Documents
Z130982
Nalgene® Griffin beakers
capacity 1,000 mL, translucent polypropylene
Synonym(s):
PP beaker, nalgene PP beaker
Select a Size
1 PKG
$110.00
$110.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 PKG
$110.00
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
$110.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
translucent polypropylene
feature
graduations
packaging
pkg of 3 ea
manufacturer/tradename
Nalgene 1201-1000
capacity
1,000 mL
General description
Low form. With silk-screened graduations, Nalgene catalog number, size and resin codes, max. use temp., and "no flame" symbol. Meet the ISO/DIS 7056 International Standards for Laboratory Plasticware Accuracy. Chemically resistant. Autoclavable. Not for use on a hot plate.
Other Notes
Browse all plastic beakers
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)