As mentioned in the PROPERTIES section, there are 6 in one pack.
Z130915
Nalgene® Griffin beakers
capacity 250 mL, transparent polymethylpentene
Synonym(s):
PMP beaker, nalgene PMP beaker
Select a Size
$132.00
Select a Size
About This Item
$132.00
Recommended Products
material
transparent polymethylpentene
feature
graduations
packaging
pkg of 6 ea
manufacturer/tradename
Nalgene 1203-0250
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
how many beaker in one pack, please?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service