Key Documents
Z126764
Standard point hypodermic needles, 304 stainless steel
gauge 13, L 2 in.
Select a Size
12 EA
$694.00
$694.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
12 EA
$694.00
About This Item
UNSPSC Code:
41122002
eCl@ss:
32070500
NACRES:
NB.03
$694.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
304 stainless steel needle
chrome-plated hub
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science
L
2 in.
gauge
13
Related Categories
General description
Hypodermic needles are precision made and designed to ensure strict adherence with U.S. Government specification GG-N-196. Points are machine ground to achieve uniform sharpness. Precision grinding, micro-bead jet blasting, and ultrasonic cleaning assure freedom from burrs and debris. All needle cannula are fabricated of type 304 stainless steel tubing which must pass rigorous quality control prior to acceptance and use.
Standard nickle plated Luer hubs comply with Luer taper specifications ISO 594. This assures an interchangeable fit mating with Luer slips and Luer locks. Contoured hub cavity facilitates cleaning.
Choose from one of the most recent versions:
Certificates of Analysis (COA)