Key Documents
CAD7932
Laboratory pipetting needle with 90° blunt ends
gauge 20, L 1 1/2 in., nickel plated hub
Select a Size
12 EA
$353.00
$353.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
12 EA
$353.00
About This Item
UNSPSC Code:
41122002
NACRES:
NB.03
$353.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
nickel plated hub
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 7932
L
1 1/2 in.
gauge
20
General description
Pipetting needles are available in a wide choice of sizes for use in laboratory procedures. All needles are made with type 304 stainless steel cannula.
Catalog numbers CAD7950 to CAD7957 needles are fitted with stainless steel hubs for special applications with corrosive materials or critical procedures. Catalog numbers CAD7930 to CAD7942 needles are fitted with standard nickel plated Luer hubs. These needles are used in conjunction with both manual and automatic volumetric dispensers, dilutors, and pipettors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)