Key Documents
Z122459
Sigma-Aldrich
Aldrich® three-neck round-bottom flask
angled, capacity 100 mL, joint: ST/NS 14/20
Synonym(s):
flask round bottom, round bottom flasks, round bottomed flask
Select a Size
1 EA
$129.00
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$129.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom flask
three-neck
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)