Z119679
Cochranes molecular models
organic/inorganic, minit, single kit with booklet
Synonym(s):
chemistry model sets, molecular model sets
Select a Size
$63.80
Select a Size
About This Item
$63.80
Recommended Products
description
Scale: 2 cm = 0.1 nanometer (1 Å), Minit 3 cm = 0.1 nanometer (1 Å), Orbit
feature
no. of atoms 240
no. of bonds 220
manufacturer/tradename
Cochranes 0073
Looking for similar products? Visit Product Comparison Guide
General description
Please see Technical Information Bulletin AL-183 for a listing of these items, available on our website at sigma-aldrich.com
ATOMS:
- 1-prong: Hydrogen, white (50), Nitrogen, blue (10), Oxygen, red (10), Fluorine, light green (10), Sulfur, yellow (2), Chlorine, green (15)
- 2-prong linear: Carbon, black (10), Nitrogen, blue (2)
- 2-prong 100°: Sulfur, yellow (10)
- 2-prong 110°: Oxygen, red (25)
- 3-prong 120°: Carbon, black (20), Nitrogen, blue (6)
- 4-prong tetrahedral: Carbon, black (30), Nitrogen, blue (10), Oxygen, red (10), Phosphorous, purple (5), Sulfur, yellow (5), Metal, silver (2)
- 6-prong octahedral: Carbon, black (2), Sulfur, yellow (1), Metal, silver (2)
- 5-prong trigonal bipyramid: Carbon, black (6), Nitrogen, blue (1), Oxygen, red (1), Phosphorous, purple (1)
- W pegs (8)
BONDS:
- Straw, green, 2.5 cm (80)
- Straw, green, 2 cm (20)
- Straw, green, 1.5 cm (80)
- Straw, red, 3 cm (12)
- Straw, red, 2 cm (12)
- Straw, mixed set/26 (1)
- St v flexible white, 5 cm (4)
- St v flexible white, 2.5 cm (6)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Use our handy VSEPR chart to find the 3-D geometric VSEPR shapes of molecules and ions and learn about VSEPR theory and shapes.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service