Key Documents
Z115738
Sigma-Aldrich
Aldrich® rotary evaporator cleaning adapter
joint: ST/NS 24/40
Select a Size
1 EA
$109.00
$109.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$109.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$109.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
Related Categories
General description
For fast and easy cleaning of evaporator condenser assembly.
Evaporator Cleaning Technique
1. Attach the cleaning adapter to a rotary evaporator.
2. Plug the end of the cleaning adapter.
3. Pull solvent into the system through the feed tube.
4. Quickly release the vacuum.
5. Solvent is sprayed throughout the entire assembly cleaning it quickly and efficiently.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)