Z115126
Vials, screw top V-Vials® with open-top cap
volume 0.3 mL, 13-425 thread, diam. × H 13 mm × 35 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PTFE/silicone cap liner
black phenolic cap
clear glass vial
feature
closure type black phenolic hole cap
closure type screw top vial
diam. × H
13 mm × 35 mm
volume
0.3 mL
fitting
13-425 thread
Looking for similar products? Visit Product Comparison Guide
General description
Graduated. Autoclavable borosilicate USP Type 1 glass. PTFE faced rubber septum.
Footnote
The glass in Wheaton V-Vials meets the USP and EP Type I glass requirements for the arsenic and powdered glass tests.
Legal Information
V-Vial is a registered trademark of Wheaton Industries, Inc.
related product
Product No.
Description
Pricing
replacement part
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service