Skip to Content
Merck
All Photos(1)

Key Documents

23226

Magnetic Stir Bar

for use with For 0.1-2.0mL vessels, pkg of 6 ea, PTFE

Synonym(s):

STIRRING BARS 0.3ML TO 2ML M.V. PK6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103806
Pricing and availability is not currently available.

material

PTFE
triangular PTFE-coated

packaging

pkg of 6 ea

compatibility

for use with For 0.1-2.0mL vessels

Looking for similar products? Visit Product Comparison Guide

Application

Magnetic stir bar may be used for stirring samples in the stir-bar sorptive extraction (SBSE) of volatile components of malt whisky, analyzed using gas chromatography coupled with mass spectrometry (GC-MS).[1]

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of volatiles of malt whisky by solid-phase microextraction and stir bar sorptive extraction.
Demyttenaere RCJ.
Journal of Chromatography A, 985(1-2), 221-232 (2003)

Articles

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Questions

  1. Welche Dimensionen hat der Rührfisch? 

    1 answer
    1. The total length of this stir vane is 0.373 inches, the total width is 0.218 inches. See the image below to review the supplier drawing for this item.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service