Key Documents
Z103314
Sigma-Aldrich
Aldrich® round-bottom flask with septum-inlet
capacity 100 mL, joint: ST/NS 14/20
Synonym(s):
bottom flask, flask round, round flask
Select a Size
1 EA
$113.00
$113.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$113.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$113.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom flask
two-neck
Joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
General description
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)