Key Documents
Z101230
Sigma-Aldrich
Aldrich® round-bottom flask with septum-inlet
capacity 100 mL, joint: ST/NS 19/22
Synonym(s):
bottom flask, flask round, round flask
Select a Size
1 EA
$157.00
$157.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$157.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$157.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
round bottom flask
single-neck
Joint
joint: ST/NS19/22
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
100 mL
General description
With ground-glass joint. Use septum Z565695 or Z565679 (see Septa section).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)