Skip to Content
Merck
All Photos(1)

Key Documents

WHA67245000

Whatman® Polydisc SPF in-line filter for aqueous solutions

pore size 1.0 μm, sterile

Synonym(s):

Z754277

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104922
NACRES:
NB.24
Pricing and availability is not currently available.

material

glass microfiber membrane (with glass microfiber)
polypropylene housing

sterility

sterile

packaging

pk of 10

manufacturer/tradename

Whatman 6724-5000
Whatman Article No. 28420224 (US reference)

diam.

50 mm

pore size

1.0 μm

General description

Whatman Polydisc SPF disc filters from Cytiva contain a glass microfiber (GMF) and polyethersulfone (PES) stack designed to purify serum and other hard-to-filter solutions.

Whatman membranes from Cytiva′s business offer a wide range of high-throughput, in-line options. Explore the Polydisc line to find the right disc filters for your needs.

Filtering serum requires removing proteins, lipids, salts, and other cell debris. This range of particulate matter is effectively handled with multi-layer prefilters to facilitate downstream work and to avoid clogging later serum filters.

Polydisc SPF stacks a high-flow, hydrophilic PES membrane with a high particle-loading GMF filter to clean out particulates from serum and reduce stress on the final-stage serum filters.

Features and Benefits

  • High-throughput, in-line prefilters for use upstream of serum filters
  • GMF prefilter captures large particles and cell debris while PES stack removes remaining particles and bacteria larger than 1 µm
  • Designed to extend the life of downstream serum filters
  • Effective for microbiology and tissue culture, immunoassays, virology, and diagnostic controls
  • 6 to 10 mm i.d. hose connection

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service