Skip to Content
Merck
All Photos(2)

Key Documents

W800111

Sigma-Aldrich

cis-3-Hexenyl methyl carbonate

stabilized, ≥98%, FG

Synonym(s):

(Z)-3-hexen-1-yl methyl carbonate

Sign Into View Organizational & Contract Pricing

Select a Size

W800111-SAMPLE-K
$172.00
100 G
$222.00
1 KG
$650.00

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W800111-SAMPLE-K
$172.00
100 G
$222.00
1 KG
$650.00

About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.838
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
food allergen:
no known allergens

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

contains

α-tocopherol synthetic

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; green; floral; waxy; violet

SMILES string

O(CC\C=C/CC)C(=O)OC

InChI

1S/C8H14O3/c1-3-4-5-6-7-11-8(9)10-2/h4-5H,3,6-7H2,1-2H3/b5-4-

InChI key

BLOXMGXSDAAJGX-PLNGDYQASA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service