Skip to Content
Merck
All Photos(1)

Key Documents

347736

Sigma-Aldrich

4-tert-Butylcyclohexyl acetate, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$58.20

$58.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$58.20

About This Item

Linear Formula:
CH3CO2C6H10C(CH3)3
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$58.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.452 (lit.)

bp

228-230 °C/25 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(=O)OC1CCC(CC1)C(C)(C)C

InChI

1S/C12H22O2/c1-9(13)14-11-7-5-10(6-8-11)12(2,3)4/h10-11H,5-8H2,1-4H3

InChI key

MBZRJSQZCBXRGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-tert-Butylcyclohexyl acetate has been evaluated as a fragrance ingredient and its toxicologic and dermatological analysis has been reported.[1] Cis and trans forms of 4-tert-butylcyclohexyl acetate are widely used as a perfume for cosmetics including soaps.[2]

Application

4-tert-Butylcyclohexyl acetate may be used in the preparation of high loading fragrance encapsulation based on polymer-blend of ethylcellulose, hydroxypropyl methylcellulose and poly(vinyl alcohol).[3]

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Facile Stereoselective Biocatalytic Route to the Precursor of Woody Acetate.
Brown G, et al.
Organic Process Research & Development, 15(5), 1036-1039 (2011)
Aurapan Sansukcharearnpon et al.
International journal of pharmaceutics, 391(1-2), 267-273 (2010-02-23)
The six fragrances, camphor, citronellal, eucalyptol, limonene, menthol and 4-tert-butylcyclohexyl acetate, which represent different chemical functionalities, were encapsulated with a polymer-blend of ethylcellulose (EC), hydroxypropyl methylcellulose (HPMC) and poly(vinyl alcohol) (PV(OH)) using solvent displacement (ethanol displaced by water). The process
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46 Suppl 12, S36-S41 (2008-10-10)
A toxicologic and dermatologic review of 4-tert-butylcyclohexyl acetate when used as a fragrance ingredient is presented.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service