Skip to Content
Merck
All Photos(1)

Key Documents

W490355

Sigma-Aldrich

Ethyl 3-methyl-2-oxopentanoate

≥95%

Sign Into View Organizational & Contract Pricing

Select a Size

W490355-SAMPLE-K
$76.50
100 G
$240.00
500 G
$759.00

$76.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
W490355-SAMPLE-K
$76.50
100 G
$240.00
500 G
$759.00

About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
FEMA Number:
4903
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
food allergen:
no known allergens

$76.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥95%

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; woody; walnut

SMILES string

O(CC)C(=O)C(=O)C(CC)C

InChI

1S/C8H14O3/c1-4-6(3)7(9)8(10)11-5-2/h6H,4-5H2,1-3H3

InChI key

HCDYSXWWIWPQEN-UHFFFAOYSA-N

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.8 °F

Flash Point(C)

77.11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service