Skip to Content
Merck
All Photos(2)

Key Documents

W489811

Sigma-Aldrich

(E)-5-Octenal

≥95%, stabilized

Synonym(s):

Pomelo aldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

W489811-SAMPLE-K
$76.50
25 G
$243.00
100 G
$449.00

$76.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W489811-SAMPLE-K
$76.50
25 G
$243.00
100 G
$449.00

About This Item

Empirical Formula (Hill Notation):
C8H14O
CAS Number:
Molecular Weight:
126.20
FEMA Number:
4898
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
food allergen:
no known allergens

$76.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 117

Assay

≥95%

contains

alpha-tocopherol as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grapefruit; citrus

SMILES string

O=CCCC\C=C\CC

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h3-4,8H,2,5-7H2,1H3/b4-3+

InChI key

ZUSUVEKHEZURSD-ONEGZZNKSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F

Flash Point(C)

56 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service