Skip to Content
Merck
All Photos(1)

Key Documents

W478620

Sigma-Aldrich

2-Hexyl-2-decenal

natural (US), mixture of cis and trans, ≥95%, stabilized

Sign Into View Organizational & Contract Pricing

Select a Size

W478620-SAMPLE-K
$52.00
100 G
$171.20
1 KG
$700.00

$52.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W478620-SAMPLE-K
$52.00
100 G
$171.20
1 KG
$700.00

About This Item

Empirical Formula (Hill Notation):
C16H30O
CAS Number:
Molecular Weight:
238.41
FEMA Number:
4786
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Halal
Kosher
natural (US)
biological source:
botanical
food allergen:
no known allergens

$52.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

Halal
Kosher
natural (US)

reg. compliance

FDA 21 CFR 117

Assay

≥95%

contains

BHA as stabilizer

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; waxy

SMILES string

O=CC(=CCCCCCCC)CCCCCC

InChI

1S/C16H30O/c1-3-5-7-9-10-12-14-16(15-17)13-11-8-6-4-2/h14-15H,3-13H2,1-2H3

InChI key

RSBRHLFCWKXUSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service