Skip to Content
Merck
All Photos(1)

Key Documents

W434100

Sigma-Aldrich

Heptyl heptanoate

natural (US), 98%

Synonym(s):

Heptanoic acid, heptyl ester, Heptyl capronate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H28O2
CAS Number:
Molecular Weight:
228.37
FEMA Number:
4341
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Organoleptic:
grassy; green; fruity
grade:
Halal
Kosher
natural (US)
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

Halal
Kosher
natural (US)

reg. compliance

FDA 21 CFR 117

Assay

98%

refractive index

n20/D 1.431

density

0.858 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

grassy; green; fruity

SMILES string

O=C(CCCCCC)OCCCCCCC

InChI

1S/C14H28O2/c1-3-5-7-9-11-13-16-14(15)12-10-8-6-4-2/h3-13H2,1-2H3

InChI key

QOIIBPAJVWFEPE-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service