Skip to Content
Merck
All Photos(1)

Key Documents

W474625

Sigma-Aldrich

3,5-Undecadien-2-one

10% solution in EtOH, natural (US)

Sign Into View Organizational & Contract Pricing

Select a Size

W474625-SAMPLE-K
$58.30
100 G
$233.00
500 G
$758.00

$58.30


Estimated to ship onMay 28, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W474625-SAMPLE-K
$58.30
100 G
$233.00
500 G
$758.00

About This Item

Empirical Formula (Hill Notation):
C11H18O
CAS Number:
Molecular Weight:
166.26
FEMA Number:
4746
UNSPSC Code:
12164502
NACRES:
NA.21
grade:
Kosher
natural (US)
food allergen:
no known allergens

$58.30


Estimated to ship onMay 28, 2025


Request a Bulk OrderRequest more information

grade

Kosher
natural (US)

Quality Level

reg. compliance

FDA 21 CFR 117

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cucumber; fatty; green

SMILES string

O=C(C)\C=C\C=C\CCCCC

InChI

1S/C11H18O/c1-3-4-5-6-7-8-9-10-11(2)12/h7-10H,3-6H2,1-2H3/b8-7+,10-9+

InChI key

RCFOEQFXUGQOMT-XBLVEGMJSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service