Skip to Content
Merck
All Photos(3)

Documents

W240115

Sigma-Aldrich

δ-Dodecalactone

greener alternative

mixture of isomers, natural, ≥98%, FG

Synonym(s):

(±)-5-Dodecanolide, (±)-δ-Heptyl-δ-valerolactone, (±)-6-Heptyltetrahydro-2H-pyran-2-one, 5-Hydroxydodecanoic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
FEMA Number:
2401
Beilstein:
1282749
EC Number:
Council of Europe no.:
624
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.008
NACRES:
NA.21

biological source

Cocos nucifera (oil)

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.460 (lit.)

bp

140-141 °C/1 mmHg (lit.)

mp

−12 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

tree nuts

fragrance allergen

no known allergens

greener alternative category

Organoleptic

butter; cheese; coconut; creamy; oily; peach

SMILES string

CCCCCCCC1CCCC(=O)O1

InChI

1S/C12H22O2/c1-2-3-4-5-6-8-11-9-7-10-12(13)14-11/h11H,2-10H2,1H3

InChI key

QRPLZGZHJABGRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Other Notes

Natural occurrence: Blue cheese, heated butter, pork fat, rum, sherry and cheeses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service