Skip to Content
Merck
All Photos(1)

Key Documents

W376418

Sigma-Aldrich

Menthalactone

≥99%

Synonym(s):

mint lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22
Beilstein:
1343029
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
10.036
NACRES:
NA.21

biological source

synthetic

grade

Halal

Agency

meets purity specifications of JECFA

Assay

≥99%

optical activity

[α]20/D −35°, neat

refractive index

n20/D 1.5 (lit.)

bp

87-89 °C/25 mmHg (lit.)

density

1.058 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coconut; creamy; spearmint; sweet; tobacco

SMILES string

CC1CC=C2C(C)C(=O)OC2C1

InChI

1S/C10H14O2/c1-6-3-4-8-7(2)10(11)12-9(8)5-6/h4,6-7,9H,3,5H2,1-2H3

InChI key

NOSVNHVKMHWUAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Menthalactone occurs naturally in mint. It may be contributing to its analgesic activity.

Biochem/physiol Actions

Odor at 10%
Taste at 20 ppm

Other Notes

Natural occurrence: Peppermint and penny royal oil, Mentha arvensis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Menthalactone, a new analgesic from Mentha cordifolia Opiz. leaves.
Villase?or IM & Sanchez AC.
Zeitschrift fur Naturforschung C, 64(11-12), 809-812 (2009)
Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 30, 48-48 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service