Skip to Content
Merck
All Photos(1)

Key Documents

W366005

Sigma-Aldrich

9-Decenoic acid

≥95%, stabilized, FG

Synonym(s):

9-decylenic acid, Caproleic acid

Sign Into View Organizational & Contract Pricing

Select a Size

W366005-SAMPLE-K
$56.20
25 G
$184.00
100 G
$498.00
1 KG
$3,740.00

$56.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W366005-SAMPLE-K
$56.20
25 G
$184.00
100 G
$498.00
1 KG
$3,740.00

About This Item

Linear Formula:
H2C=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
170.25
FEMA Number:
3660
EC Number:
Council of Europe no.:
10090
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.065
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$56.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥95%

contains

<0.10% α-tocopherol, synthetic as stabilizer

composition

decenoic acid, ≥95% (mixture of isomers)

refractive index

n20/D 1.447 (lit.)

bp

270 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

milk; fruity; waxy

SMILES string

OC(=O)CCCCCCCC=C

InChI

1S/C10H18O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2H,1,3-9H2,(H,11,12)

InChI key

KHAVLLBUVKBTBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C E Serna-Jiménez et al.
International journal of pharmaceutics, 492(1-2), 223-232 (2015-07-22)
The aim of this study was to develop and evaluate a transdermal delivery system of pizotifen malate. Pizotifen is frequently used in the preventive treatment of migraine, but is also indicated in eating disorders. In the course of the project

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service