Skip to Content
Merck
All Photos(1)

Key Documents

W347108

Sigma-Aldrich

(−)-Ambroxide

≥99%

Synonym(s):

1,5,5,9-Tetramethyl-13-oxatricyclo[8.3.0.04,9]tridecane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H28O
CAS Number:
Molecular Weight:
236.39
FEMA Number:
3471
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
biological source:
synthetic

biological source

synthetic

Assay

≥99%

optical activity

[α]20/D −29.5°, c = 1 in toluene

mp

74-76 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

woody; pine

SMILES string

CC1(C)CCC[C@@]2(C)[C@H]1CC[C@@]3(C)OCC[C@H]23

InChI

1S/C16H28O/c1-14(2)8-5-9-15(3)12(14)6-10-16(4)13(15)7-11-17-16/h12-13H,5-11H2,1-4H3/t12-,13+,15-,16+/m0/s1

InChI key

YPZUZOLGGMJZJO-LQKXBSAESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asma Nasib et al.
Journal of natural products, 69(6), 957-959 (2006-06-24)
Biotransformation of (-)-ambrox (1) with cell suspension cultures of Actinidia deliciosa (Kiwifruit) yielded the regio- and stereospecific oxygenated products 3-oxoambrox (2), 3beta-hydroxyambrox (3), 1alpha-hydroxyambrox (4), 3beta,6beta-dihydroxyambrox (5), 1alpha,6beta-dihydroxyambrox (6), and 1alpha,3beta-dihydroxyambrox (7). Metabolites 6 and 7 were found to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service