Skip to Content
Merck
All Photos(1)

Key Documents

357995

Sigma-Aldrich

Sclareol

98%

Synonym(s):

Labd-14-ene-8,13-diol, (1R,2R,8aS)-Decahydro-1-(3-hydroxy-3-methyl-4-pentenyl)-2,5,5,8a-tetramethyl-2-naphthol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$108.00

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$108.00

About This Item

Empirical Formula (Hill Notation):
C20H36O2
CAS Number:
Molecular Weight:
308.50
Beilstein:
2054148
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$108.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

optical activity

[α]25/D −13°, c = 4 in carbon tetrachloride

bp

218-220 °C/19 mmHg (lit.)

mp

95-100 °C (lit.)

functional group

hydroxyl

SMILES string

CC1(C)CCC[C@@]2(C)[C@H]1CC[C@@](C)(O)[C@@H]2CC[C@@](C)(O)C=C

InChI

1S/C20H36O2/c1-7-18(4,21)13-9-16-19(5)12-8-11-17(2,3)15(19)10-14-20(16,6)22/h7,15-16,21-22H,1,8-14H2,2-6H3/t15-,16+,18-,19-,20+/m0/s1

InChI key

XVULBTBTFGYVRC-HHUCQEJWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sclareol, a diterpene compound with a labdane skeleton, is mainly used as a raw material in the fragrance industry.[1] It shows potent cytotoxic and cytostatic effect in human leukemic cell lines.[2]

Application

Sclareol may be used as a starting material in the synthesis of ambergris fragrance chemicals such as ambraoxide, ambrox, methylambraoxide, ambracetal, ambraketal and epiambraketal.[3][4] It may also be used in the synthesis of (+)-galanolactone, (-)-8-epi-galanolactone and (+)-labdienedial.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Discovery and functional characterization of two diterpene synthases for sclareol biosynthesis in Salvia sclarea (L.) and their relevance for perfume manufacture.
Caniard A, et al.
BMC plant biology, 12(1), 119-119 (2012)
A short efficient synthesis of ambraketal (four steps) and epiambraketal (five steps) from Sclareol.
Martres P, et al.
Tetrahedron Letters, 35(1), 97-98 (1994)
The effect of sclareol on growth and cell cycle progression of human leukemic cell lines.
Dimas K, et al.
Leukemia Research, 23(3), 217-234 (1999)
Synthesis of ambergris fragrance chemicals from sclareol, involving palladium catalysed key steps.
Coste-Maniere IC, et al.
Tetrahedron Letters, 29(9), 1017-1020 (1988)
Conversion of sclareol into (+)-galanolactone and (+)-labdienedial.
Jung M, et al.
Tetrahedron Letters, 38(16), 2871-2874 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service