Skip to Content
Merck
All Photos(1)

Key Documents

W329002

Sigma-Aldrich

3-Hexanone

≥97%

Synonym(s):

Ethyl propyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

W329002-SAMPLE
$54.60
25 G
$105.00
1 KG
$829.00

$54.60


Estimated to ship onMarch 11, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W329002-SAMPLE
$54.60
25 G
$105.00
1 KG
$829.00

About This Item

Linear Formula:
CH3CH2CH2COC2H5
CAS Number:
Molecular Weight:
100.16
FEMA Number:
3290
Beilstein:
1738025
EC Number:
Council of Europe no.:
4125
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.096
NACRES:
NA.21

$54.60


Estimated to ship onMarch 11, 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

Assay

≥97%

refractive index

n20/D 1.400 (lit.)

bp

123 °C (lit.)

density

0.815 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; waxy; sweet

SMILES string

CCCC(=O)CC

InChI

1S/C6H12O/c1-3-5-6(7)4-2/h3-5H2,1-2H3

InChI key

PFCHFHIRKBAQGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Hexanone has been reported as one of the volatile organic compounds (VOCs) in pandan leaves[1], strawberry[2] and acerola fruit.[3]

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F

Flash Point(C)

14 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile composition of pandan leaves (Pandanus amaryllifolius).
Flavor chemistry of ethnic foods, 105-109 (1999)
Metabolism of natural volatile compounds by strawberry fruit.
Hamilton-Kemp TR, et al.
Journal of Agricultural and Food Chemistry, 44(9), 2802-2805 (1996)
Volatile flavor constituents of acerola (Malpighia emarginata DC.) fruit.
Pino JA & Marbot R.
Journal of Agricultural and Food Chemistry, 49(12), 5880-5882 (2001)
Blas Agut et al.
The New phytologist, 207(3), 790-804 (2015-03-17)
The citrus rootstocks sour orange and Cleopatra mandarin display differential resistance against Tetranychus urticae. Sour orange plants support reduced oviposition, growth rates and damage compared with Cleopatra mandarin plants. Jasmonic acid signalling and flavonoid accumulation have been revealed as key
Profile of volatile metabolites in human urine.
A Zlatkis et al.
Clinical chemistry, 17(7), 592-594 (1971-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service