Skip to Content
Merck
All Photos(2)

Key Documents

108707

Sigma-Aldrich

2-Methyl-3-pentanone

97%

Synonym(s):

Ethyl isopropyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$84.20
25 G
$248.00

$84.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$84.20
25 G
$248.00

About This Item

Linear Formula:
(CH3)2CHCOC2H5
CAS Number:
Molecular Weight:
100.16
Beilstein:
1698849
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39021212
PubChem Substance ID:
NACRES:
NA.22

$84.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.397 (lit.)

bp

113 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCC(=O)C(C)C

InChI

1S/C6H12O/c1-4-6(7)5(2)3/h5H,4H2,1-3H3

InChI key

HYTRYEXINDDXJK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-3-pentanone may be used to study the stereochemistry of ketone enolization by lithium 2, 2, 6, 6-tetramethylpiperidide[1].

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of lithium salts on the stereochemistry of ketone enolization by lithium 2, 2, 6, 6-tetramethylpiperidide (LiTMP). A convenient method for highly E-selective enolate formation.
Hall PL, et al.
Journal of the American Chemical Society, 113(25), 9571-9574 (1991)
Catherine E Garner et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 21(8), 1675-1688 (2007-02-23)
Little is known about the volatile organic compounds (VOCs) in feces and their potential health consequences. Patients and healthcare professionals have observed that feces often smell abnormal during gastrointestinal disease. The aim of this work was to define the volatiles
Profile of volatile metabolites in human urine.
A Zlatkis et al.
Clinical chemistry, 17(7), 592-594 (1971-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service