Skip to Content
Merck
All Photos(1)

Key Documents

W310808

Sigma-Aldrich

Vanillin acetate

≥98%, FG

Synonym(s):

4-Formyl-2-methoxyphenyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

W310808-SAMPLE-K
$54.60
100 G
$70.00
1 KG
$240.00
5 KG
$1,020.00

$54.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
W310808-SAMPLE-K
$54.60
100 G
$70.00
1 KG
$240.00
5 KG
$1,020.00

About This Item

Linear Formula:
CH3CO2C6H3-4-(CHO)-2-OCH3
CAS Number:
Molecular Weight:
194.18
FEMA Number:
3108
EC Number:
Council of Europe no.:
225
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.035
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$54.60

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 30, 2025


Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

Assay

≥98%

mp

77-79 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

creamy; sweet; vanilla

SMILES string

COc1cc(C=O)ccc1OC(C)=O

InChI

1S/C10H10O4/c1-7(12)14-9-4-3-8(6-11)5-10(9)13-2/h3-6H,1-2H3

InChI key

PZSJOBKRSVRODF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Vanilla Regulations – The Law and the Labeling of the World’s Most Popular Flavor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service