Skip to Content
Merck
All Photos(1)

Key Documents

W304603

Sigma-Aldrich

Terpinolene

≥95%, FG

Synonym(s):

4-Isopropylidene-1-methylcyclohexene, p-Menth-1,4(8)-diene, p-Meth-1-en-8-yl-formate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
3046
Beilstein:
1851203
EC Number:
Council of Europe no.:
2115
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.005
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU 1334/2008 & 872/2012

vapor density

~4.7 (vs air)

vapor pressure

~0.5 mmHg ( 20 °C)

Assay

≥95%

refractive index

n20/D 1.489 (lit.)

bp

184-185 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fresh; woody; citrus; pine; sweet

SMILES string

C\C(C)=C1/CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4H,5-7H2,1-3H3

InChI key

MOYAFQVGZZPNRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terpinolene is a volatile monoterpene that occurs naturally in sage, rosemary and carrot.

Application


  • Based on HPLC and HS-GC-IMS Techniques, the Changes in the Internal Chemical Components of Schisandra chinensis (Turcz.) Baill. Fruit at Different Harvesting Periods Were Analyzed.: This study utilized high-performance liquid chromatography and headspace gas chromatography-ion mobility spectrometry to examine changes in internal chemical components, including terpinolene, of Schisandra chinensis during various harvest periods, offering insights into optimal harvest timing for maximum chemical efficacy (Sun et al., 2024).

  • Exogenous methyl jasmonate treatment induced the transcriptional responses and accumulation of volatile terpenoids in Oenanthe javanica (Blume) DC.: Research focused on the effect of methyl jasmonate on the accumulation of volatile terpenoids, such as terpinolene, in Oenanthe javanica, demonstrating enhanced production of these compounds which are critical for plant defense mechanisms (Feng et al., 2024).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - Seta closed cup

Flash Point(C)

61 °C - Seta closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carrot flavor: effects of genotype, growing conditions, storage, and processing.
Simon PW.
Evaluation of quality of fruits and vegetables, 315-328 (1985)
Terpinolene, a component of herbal sage, downregulates AKT1 expression in K562 cells.
Okumura N, et al
Oncology Letters, 3(2), 321-324 (2012)
Yan Ma et al.
Physical chemistry chemical physics : PCCP, 11(21), 4198-4209 (2009-05-22)
Gas-phase ozonolysis of terpinolene was studied in static chamber experiments using gas chromatography coupled to mass spectrometric and flame ionisation detection to separate and detect products. Two isomers of C(7)-diacids and three isomers of C(7)-aldehydic acids were identified in the
Barbara Conti et al.
Parasitology research, 110(5), 2013-2021 (2011-12-14)
Lamiaceae have traditionally been used in developing countries for their insecticidal and repellent properties against several insect species. In our research, the essential oil (EO) extracted from fresh leaves of Hyptis suaveolens (Lamiaceae), and its main constituents were evaluated for
John H Borden et al.
Journal of economic entomology, 101(4), 1266-1275 (2008-09-05)
The superiority of the host monoterpene myrcene as a synergist for trans-verbenol and exo-brevicomin, aggregation pheromone components of the mountain pine beetle, Dendroctonus ponderosae Hopkins (Coleoptera: Curculionidae), suggests that the ancestral host of the mountain pine beetle is a pine

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service