Skip to Content
Merck
All Photos(4)

Key Documents

W276212

Sigma-Aldrich

Myrcene

greener alternative

sum of isomers, ≥90%, natural, FG

Synonym(s):

β-Myrcene, 7-Methyl-3-methylene-1,6-octadiene

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$114.00
W276212-SAMPLE-K
$169.00
1 KG
$596.00

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
100 G
$114.00
W276212-SAMPLE-K
$169.00
1 KG
$596.00

About This Item

Linear Formula:
H2C=CHC(=CH2)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
136.23
FEMA Number:
2762
Beilstein:
1719990
EC Number:
MDL number:
UNSPSC Code:
12352212
Flavis number:
1.008
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR part: 172.510

vapor density

4.7 (vs air)

vapor pressure

~7 mmHg ( 20 °C)

Assay

≥90%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.469 (lit.)

bp

167 °C (lit.)

density

0.791 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

limonene (sum of D, L and DL)

greener alternative category

Organoleptic

balsamic; herbaceous; woody

storage temp.

2-8°C

SMILES string

C\C(C)=C/CCC(=C)C=C

InChI

1S/C10H16/c1-5-10(4)8-6-7-9(2)3/h5,7H,1,4,6,8H2,2-3H3

InChI key

UAHWPYUMFXYFJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Selective Reactivity of Myrcene for Vat Photopolymerization 3D Printing and Postfabrication Surface Modification.: This research highlights the selective reactivity of myrcene in vat photopolymerization for 3D printing, demonstrating its potential in creating functionalized surfaces for biomedical applications (Weems et al., 2020, Biomacromolecules).

  • Inverse Vulcanization of Sulfur using Natural Dienes as Sustainable Materials for Lithium-Sulfur Batteries.: This article explores the use of myrcene and other natural dienes in the inverse vulcanization of sulfur, promoting sustainable materials for lithium-sulfur batteries (Gomez et al., 2016, ChemSusChem).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tai-Ti Liu et al.
Insects, 11(1) (2020-01-02)
Bacopa caroliniana (BC) is a perennial creeping herb and popular aquarium plant. This plant is easily cultivated; consequently, it has the potential to be a raw material which is readily available for mass production if it contains useful bioactive substances.
R Tognon et al.
Bulletin of entomological research, 110(5), 638-644 (2020-05-10)
Telenomus podisi Ashmead (Hymenoptera: Scelionidae) is the most important egg parasitoid of Euschistus heros (Fabricius) (Hemiptera: Pentatomidae), and its successful parasitism is related to their searching ability to find suitable hosts under a complex chemical environment using host-reliable cues. Thus
Bipana Paudel Timilsena et al.
Plant, cell & environment, 43(3), 787-800 (2019-11-24)
Plants produce species-specific herbivore-induced plant volatiles (HIPVs) after damage. We tested the hypothesis that herbivore-specific HIPVs prime neighboring plants to induce defenses specific to the priming herbivore. Since Manduca sexta (specialist) and Heliothis virescens (generalist) herbivory induced unique HIPV profiles
Yaoyao Peng et al.
Food research international (Ottawa, Ont.), 129, 108873-108873 (2020-02-11)
The phenolic compounds and aroma active compounds in feijoa (Acca sellowiana (O.Berg) Burret) juice from four New Zealand grown cultivars (Apollo, Unique, Opal Star, and Wiki Tu) were investigated. A high total phenolic content (maximum 1.89 mg GAE/mL juice) and significant
Paroma Mitra et al.
Pest management science, 77(1), 285-299 (2020-07-23)
The viviparous aphid Aphis craccivora Koch (Hemiptera: Aphididae) is a serious threat to the crop yield of Lathyrus sativus L. (Fabaceae), commonly known as grass pea. The synthetic insecticides applied to control this insect pest are not safe for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service