Skip to Content
Merck
All Photos(1)

Key Documents

W231304

Sigma-Aldrich

Citronellyl isobutyrate

≥92%, FCC

Sign Into View Organizational & Contract Pricing

Select a Size

W231304-SAMPLE-K
$59.80
1 KG
$160.00

$59.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W231304-SAMPLE-K
$59.80
1 KG
$160.00

About This Item

Linear Formula:
(CH3)2CHCO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
226.36
FEMA Number:
2313
EC Number:
Council of Europe no.:
296
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.421
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens

$59.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥92%

refractive index

n20/D 1.442 (lit.)

bp

253 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

citrus; fruity; tropical; sweet

SMILES string

CC(CCOC(=O)C(C)C)CC\C=C(\C)C

InChI

1S/C14H26O2/c1-11(2)7-6-8-13(5)9-10-16-14(15)12(3)4/h7,12-13H,6,8-10H2,1-5H3

InChI key

ZGPPERKMXSGYRK-UHFFFAOYSA-N

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service