Skip to Content
Merck
All Photos(1)

Key Documents

W231200

Sigma-Aldrich

Citronellyl butyrate

natural (US), ≥85%, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CO2CH2CH2CH(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
226.36
FEMA Number:
2312
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.049
NACRES:
NA.21
grade:
FG
Kosher
natural (US)
food allergen:
no known allergens

grade

FG
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥85%

refractive index

n20/D 1.445 (lit.)

bp

245 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; fruity; citrus; floral; sweet

SMILES string

CCCC(=O)OCCC(C)CC\C=C(\C)C

InChI

1S/C14H26O2/c1-5-7-14(15)16-11-10-13(4)9-6-8-12(2)3/h8,13H,5-7,9-11H2,1-4H3

InChI key

XQPZQXTWYZAXAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Material may darken over time.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service