Skip to Content
Merck
All Photos(1)

Key Documents

W226718

Sigma-Aldrich

Cedar leaf oil

Synonym(s):

Methyl 2-hexynoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2267
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
biological source:
Thuja occidentalis L.
food allergen:
no known allergens

biological source

Thuja occidentalis L.

optical activity

[α]25/D -7 to -14

origin

USA origin

refractive index

n20/D 1.456-1.460 (lit.)

density

0.906-0.920 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

cedar

Preparation Note

Extraction method: steam distillation

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service