Skip to Content
Merck
All Photos(2)

Key Documents

M32100

Sigma-Aldrich

Methyl 4-bromocrotonate

85%, technical grade

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$156.00
100 G
$428.00

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$156.00
100 G
$428.00

About This Item

Linear Formula:
BrCH2CH=CHCO2CH3
CAS Number:
Molecular Weight:
179.01
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$156.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

85%

form

liquid

impurities

<15% 2(5H)-furanone

refractive index

n20/D 1.501 (lit.)

bp

83-85 °C/13 mmHg (lit.)

density

1.522 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)\C=C\CBr

InChI

1S/C5H7BrO2/c1-8-5(7)3-2-4-6/h2-3H,4H2,1H3/b3-2+

InChI key

RWIKCBHOVNDESJ-NSCUHMNNSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service