Key Documents
TEM-CF300-CU-TH
Sigma-Aldrich
Carbon Film Supported Copper Square Mesh
Thick, size 300 mesh, box of 50
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
size:
300 mesh
packaging:
box of 50
Product TEM-CF300-CU-TH is currently not available for purchase. Contact Technical Service
packaging
box of 50
size
300 mesh
General description
A 10 nm thick film of pure carbon supported standard squre mesh grids. Copper grid, 300 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)