Skip to Content
Merck
All Photos(1)

Key Documents

T70203

Sigma-Aldrich

Trimethoxyboroxine

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$78.80

$78.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$78.80

About This Item

Empirical Formula (Hill Notation):
C3H9B3O6
CAS Number:
Molecular Weight:
173.53
Beilstein:
1709158
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$78.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

30 mmHg ( 25 °C)

Assay

95%

refractive index

n20/D 1.4 (lit.)

bp

130 °C (lit.)

mp

10 °C (lit.)

density

1.195 g/mL at 25 °C (lit.)

SMILES string

COb1ob(OC)ob(OC)o1

InChI

1S/C3H9B3O6/c1-7-4-10-5(8-2)12-6(9-3)11-4/h1-3H3

InChI key

ZFMOJHVRFMOIGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service