Skip to Content
Merck
All Photos(1)

Key Documents

T6008

Sigma-Aldrich

Tetrabutyltin

technical grade, 93%

Synonym(s):

Tin tetrabutyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4Sn
CAS Number:
Molecular Weight:
347.17
Beilstein:
3648237
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

93%

form

liquid

reaction suitability

core: tin

refractive index

n20/D 1.473 (lit.)

bp

127-145 °C/10 mmHg (lit.)

mp

−97 °C (lit.)

density

1.057 g/mL at 25 °C (lit.)

SMILES string

CCCC[Sn](CCCC)(CCCC)CCCC

InChI

1S/4C4H9.Sn/c4*1-3-4-2;/h4*1,3-4H2,2H3;

InChI key

AFCAKJKUYFLYFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

thymus,spleen

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

231.8 - 239.0 °F - closed cup

Flash Point(C)

111 - 115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Federica Cacciatore et al.
Ecotoxicology and environmental safety, 147, 688-698 (2017-09-22)
The aim of this study was to compare imposex and butyltin compounds (BTs) data, collected before and after the organotin ban in 2008, in order to assess temporal and spatial variation of the phenomenon, the decline of BT contamination, and
Xiaoyu Xiao et al.
Ecotoxicology and environmental safety, 192, 110289-110289 (2020-02-18)
Comparatively limited knowledge is known about the accumulation processes of tributyltin (TBT) and triphenyltin (TPT) in fish and aquatic plant in the freshwater environment, which has hindered a full understanding of their bioaccumulation potential and ecological risks. In the present
Jacek A Szymura et al.
Journal of mass spectrometry : JMS, 38(8), 817-822 (2003-08-26)
A new chemometric procedure called band composition analysis (BCA) designed for the deconvolution of mass spectra of organometallics is proposed. BCA generates theoretical bands T(i), then combines them to obtain a model band M, which is finally compared with the
Hai-bing Xu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 25(3), 470-472 (2005-07-15)
MCM-41 modified with tetrabutyltin and Sn-O-MCM-41 obtained from calcination of the modified MCM-41 in pure oxygen were characterized by XRD, FTIR, UV-Vis DRS, and TG analysis. It was found that the hydrothermal stability, surface acidity, and coordinative states of tin
M Ema et al.
Journal of applied toxicology : JAT, 16(1), 71-76 (1996-01-01)
Dibutyltin dichloride (DBT), tributyltin chloride (TrBT) and tetrabutyltin (TeBT) were compared for their developmental toxicity and teratogenic potential following administration during the susceptible period for teratogenesis of TrBT. Pregnant rats were given either DBT or TrBT at a dose of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service